5-(4-Methoxyphenyl)-2-thiophenecarbohydrazide
Supplier: Apollo Scientific
Synonyms:
5-(4-Methoxyphenyl)thiophene-2-carbohydrazide
OR15414-500MG
OR15414-1G
APOSOR15414-500MGEA
546
EUR
APOSOR15414-500MG
APOSOR15414-1G
5-(4-Methoxyphenyl)-2-thiophenecarbohydrazide
5-(4-Methoxyphenyl)-2-thiophenecarbohydrazide
Formula:
C₁₂H₁₂N₂O₂S MW: 248,31 g/mol Melting Pt: 195…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200930 CAS Number: 69202-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...