1-Methyl-4-(4-nitrophenyl)piperazine 98%
Supplier: Apollo Scientific
|
Warning
|
OR15422-5G
OR15422-25G
APOSOR15422-5GEA
51
EUR
APOSOR15422-5G
APOSOR15422-25G
1-Methyl-4-(4-nitrophenyl)piperazine 98%
1-Methyl-4-(4-nitrophenyl)piperazine
Formula:
C₁₁H₁₅N₃O₂ MW: 221,26 g/mol Boiling Pt: 369,5 °C Melting Pt: 109…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00101003 CAS Number: 16155-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...