2,3-Dibromo-2-(trichloroacetyl)pyrrole

Supplier: Apollo Scientific
Warning

OR15459-1G OR15459-5G
APOSOR15459-1GEA 176 EUR
APOSOR15459-1G APOSOR15459-5G
2,3-Dibromo-2-(trichloroacetyl)pyrrole
2,3-Dibromo-2-(trichloroacetyl)pyrrole
Formula: C₆H₂Br₂Cl₃NO
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00662082
CAS Number: 50371-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR