1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR15486-5G
OR15486-1G
APOSOR15486-5GEA
357
EUR
APOSOR15486-5G
APOSOR15486-1G
1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid
1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid
Formula:
C₁₀H₈Cl₂O₂ MW: 231,0800 g/mol Melting Pt: 140…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044817 CAS Number: 84604-70-6 EINECS: 283-353-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...