1-Boc-4-(2-hydroxyethyl)piperazine 98%
Supplier: Apollo Scientific
|
Danger
|
OR1550-25G
OR1550-500MG
OR1550-5G
APOSOR1550-25GEA
192
EUR
APOSOR1550-25G
APOSOR1550-500MG
APOSOR1550-5G
1-Boc-4-(2-hydroxyethyl)piperazine 98%
1-Boc-4-(2-hydroxyethyl)piperazine
Formula:
C₁₁H₂₂N₂O₃ Boiling Pt: 114 °C/0,1mmHg Melting Pt: 39…43 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00728947 CAS Number: 77279-24-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...