2-Chloro-4-(methylthio)pyrimidine

Supplier: Apollo Scientific
Danger

OR15519-1G OR15519-250MG
APOSOR15519-1GEA 29 EUR
APOSOR15519-1G APOSOR15519-250MG
2-Chloro-4-(methylthio)pyrimidine
2-Chloro-4-(methylthio)pyrimidine
Formula: C₅H₅ClN₂S
MW: 160,6300 g/mol
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD09265490
CAS Number: 49844-93-1
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR