4-(Chloromethyl)-2-isopropylthiazole, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
OR15567-1G
OR15567-250MG
OR15567-5G
APOSOR15567-1GEA
40.8
EUR
APOSOR15567-1G
APOSOR15567-250MG
APOSOR15567-5G
4-(Chloromethyl)-2-isopropylthiazole, Technical Grade
4-(Chloromethyl)-2-isopropylthiazole
Formula:
C₇H₁₀ClNS MW: 175,68 g/mol Boiling Pt: 54…57 °C/0,07mmHg Storage Temperature: Ambient |
MDL Number:
MFCD06660126 CAS Number: 40516-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...