7,7,8,8-Tetracyanoquinodimethane 98%

Supplier: Apollo Scientific
Danger

Synonyms: 1,4-Bis(dicyanomethylene)cyclohexadiene, TCNQ, 7,7,8,8-Tetracyanoquinodimethane, 7,7,8,8-Tetracyanoquinonedimethane

OR1571-5G OR1571-25G
APOSOR1571-5GEA 69.6 EUR
APOSOR1571-5G APOSOR1571-25G
7,7,8,8-Tetracyanoquinodimethane 98%
7,7,8,8-Tetracyanoquinodimethane
Formula: C₁₂H₄N₄
MW: 204,19 g/mol
Melting Pt: 287…289 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011664
CAS Number: 1518-16-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR