1-Methylpyrrole-2-carbonyl chloride

Supplier: Apollo Scientific
Danger

OR15749-1G
APOSOR15749-1GEA 589 EUR
APOSOR15749-1G
1-Methylpyrrole-2-carbonyl chloride
1-Methylpyrrole-2-carbonyl chloride
Formula: C₆H₆ClNO
MW: 143,57 g/mol
Boiling Pt: 84 °C/15mmHg
Storage Temperature: Freezer
MDL Number: MFCD02094021
CAS Number: 26214-68-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR