1,1,3,3,6-Pentamethyl-7-nitro-5-indanamine
Supplier: Apollo Scientific
Synonyms:
5-Amino-7-nitro-1,1,3,3,6-pentamethylindane
OR15761-10G
OR15761-1G
OR15761-5G
APOSOR15761-10GEA
433
EUR
APOSOR15761-10G
APOSOR15761-1G
APOSOR15761-5G
1,1,3,3,6-Pentamethyl-7-nitro-5-indanamine
1,1,3,3,6-Pentamethyl-7-nitro-5-indanamine
Formula:
C₁₄H₂₀N₂O₂ MW: 248,33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03783538 CAS Number: 255393-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...