2-Chloro-3-methoxypropionic acid

Supplier: Apollo Scientific
Danger

OR15940-1G OR15940-5G
APOSOR15940-1GEA 90.2 EUR
APOSOR15940-1G APOSOR15940-5G
2-Chloro-3-methoxypropionic acid
2-Chloro-3-methoxypropionic acid
Formula: C₄H₇ClO₃
MW: 138,55 g/mol
Boiling Pt: 155 °C/24,8mmHg
Density: 1,3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03093624
CAS Number: 2544-05-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR