2-Pyrimidinecarboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine
OR15942-5G
OR15942-25G
APOSOR15942-5GEA
26.1
EUR
APOSOR15942-5G
APOSOR15942-25G
2-Pyrimidinecarboxylic acid 97%
2-Pyrimidinecarboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124,1 g/mol Melting Pt: 193…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00856161 CAS Number: 31519-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...