5-Pyridin-3-ylthiophene-2-carboxylic acid

Supplier: Apollo Scientific

OR16449-1G OR16449-100MG OR16449-250MG OR16449-500MG
APOSOR16449-1GEA 366 EUR
APOSOR16449-1G APOSOR16449-100MG APOSOR16449-250MG APOSOR16449-500MG
5-Pyridin-3-ylthiophene-2-carboxylic acid
5-Pyridin-3-ylthiophene-2-carboxylic acid
Formula: C₁₀H₇NO₂S
MW: 205,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06496471
CAS Number: 278803-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR