2-(Piperidin-1-yl)benzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
OR1647-10G
OR1647-5G
APOSOR1647-10GEA
161
EUR
APOSOR1647-10G
APOSOR1647-5G
2-(Piperidin-1-yl)benzaldehyde 95%
2-(Piperidin-1-yl)benzaldehyde
Formula:
C₁₂H₁₅NO MW: 189,2500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03419311 CAS Number: 34595-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...