Benzofuran-3-carboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR16562-1G
OR16562-250MG
APOSOR16562-1GEA
140
EUR
APOSOR16562-1G
APOSOR16562-250MG
Benzofuran-3-carboxylic acid 97%
Benzofuran-3-carboxylic acid
Formula:
C₉H₆O₃ MW: 162,1400 g/mol Melting Pt: 163…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD01006722 CAS Number: 26537-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...