3-Bromo-5-chloropyridine-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR16596-1G
OR16596-5G
OR16596-10G
APOSOR16596-1GEA
21.9
EUR
APOSOR16596-1G
APOSOR16596-5G
APOSOR16596-10G
3-Bromo-5-chloropyridine-2-carboxylic acid
3-Bromo-5-chloropyridine-2-carboxylic acid
Formula:
C₆H₃BrClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12827551 CAS Number: 1189513-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...