4-Chloroindole-3-carbaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloro-1H-indole-3-carbaldehyde
OR1683-5G
APOSOR1683-5GEA
126
EUR
APOSOR1683-5G
4-Chloroindole-3-carbaldehyde 98%
4-Chloroindole-3-carbaldehyde
Formula:
C₉H₆ClNO MW: 179,6030 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05864704 CAS Number: 876-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...