Imidazo[1,2-a]pyridine-6-carbohydrazide

Supplier: Apollo Scientific
Warning

Synonyms: Imidazo[1,2-a]pyridine-6-carboxylic acid hydrazide

OR16906-1G OR16906-5G OR16906-250MG
APOSOR16906-1GEA 61.1 EUR
APOSOR16906-1G APOSOR16906-5G APOSOR16906-250MG
Imidazo[1,2-a]pyridine-6-carbohydrazide
Imidazo[1,2-a]pyridine-6-carbohydrazide
Formula: C₈H₈N₄O
MW: 176,179 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD07021499
CAS Number: 886361-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR