6-Methoxy-1H-indole-3-carbaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
OR1694-1G
OR1694-5G
APOSOR1694-1GEA
64
EUR
APOSOR1694-1G
APOSOR1694-5G
6-Methoxy-1H-indole-3-carbaldehyde 98%
6-Methoxy-1H-indole-3-carbaldehyde
Formula:
C₁₀H₉NO₂ MW: 175,19 g/mol Melting Pt: 186…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03848058 CAS Number: 70555-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...