Cyclopentanecarbaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Cyclopentanecarboxaldehyde
OR16978-100G
OR16978-25G
APOSOR16978-100GEA
255
EUR
APOSOR16978-100G
APOSOR16978-25G
Cyclopentanecarbaldehyde 95%
Cyclopentanecarbaldehyde
Formula:
C₆H₁₀O MW: 98,14 g/mol Boiling Pt: 140…141 °C Density: 0,919 g/cm³ (25 °C) Flash Pt: 28 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00060798 CAS Number: 872-53-7 EINECS: 212-829-7 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...