5-Bromo-3-chloropyridine-2-carboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-3-chloropicolinic acid
OR16996-1G
OR16996-5G
OR16996-25G
APOSOR16996-1GEA
21.9
EUR
APOSOR16996-1G
APOSOR16996-5G
APOSOR16996-25G
5-Bromo-3-chloropyridine-2-carboxylic acid 97%
5-Bromo-3-chloropyridine-2-carboxylic acid
Formula:
C₆H₃BrClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12827552 CAS Number: 1189513-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...