2-Hydroxyethylurea
Supplier: Apollo Scientific
Synonyms:
1-(2-Hydroxyethyl)urea
OR17003-100G
OR17003-250G
OR17003-500G
APOSOR17003-100GEA
132
EUR
APOSOR17003-100G
APOSOR17003-250G
APOSOR17003-500G
2-Hydroxyethylurea
2-Hydroxyethylurea
Formula:
C₃H₈N₂O₂ MW: 104.11 g/mol Melting Pt: 164…169 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059080 CAS Number: 2078-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...