Trimethylsilane 99.99%
Supplier: Apollo Scientific
|
Danger
|
OR17012-25G
OR17012-5G
APOSOR17012-25GEA
0
EUR
APOSOR17012-25G
APOSOR17012-5G
Trimethylsilane 99.99%
Trimethylsilane
Formula:
C₃H₁₀Si Boiling Pt: 6,7 °C Melting Pt: –135,9 °C Density: 0,634 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039791 CAS Number: 993-07-7 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...