4-Bromo-1-indanone

Supplier: Apollo Scientific
Warning

Synonyms: 4-bromo-2,3-dihydro-1H-inden-1-one

OR17021-100G
APOSOR17021-100GEA 109 EUR
APOSOR17021-100G
4-Bromo-1-indanone
4-Bromo-1-indanone
Formula: C₉H₇BrO
MW: 211,06 g/mol
Boiling Pt: 125 °C/1,5mmHg
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01719772
CAS Number: 15115-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR