2-Chloro-4-ethylpyrimidine 98%
Supplier: Apollo Scientific
|
Warning
|
OR17114-1G
OR17114-5G
APOSOR17114-1GEA
219
EUR
APOSOR17114-1G
APOSOR17114-5G
2-Chloro-4-ethylpyrimidine 98%
2-Chloro-4-ethylpyrimidine
Formula:
C₆H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD10699651 CAS Number: 188707-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...