Benzil dioxime

Supplier: Apollo Scientific
Warning

Synonyms: Diphenylglyoxime

OR17131-100G OR17131-25G
APOSOR17131-100GEA 143 EUR
APOSOR17131-100G APOSOR17131-25G
Benzil dioxime
Benzil dioxime
Formula: C₁₄H₁₂N₂O₂
MW: 240,26 g/mol
Melting Pt: 237…240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002113
CAS Number: 23873-81-6
EINECS: 245-921-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR