3-Aminomethyltetrahydrofuran
Supplier: Apollo Scientific
|
Danger
|
OR17201-1G
OR17201-5G
APOSOR17201-1GEA
21.9
EUR
APOSOR17201-1G
APOSOR17201-5G
3-Aminomethyltetrahydrofuran
3-Aminomethyltetrahydrofuran
Formula:
C₅H₁₁NO Boiling Pt: 65 °C Density: 0,992 g/cm³ (25 °C) Flash Pt: 58,9 °C Storage Temperature: Ambient |
MDL Number:
MFCD08234925 CAS Number: 165253-31-6 UN: 2924 ADR: 3+8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...