5-Bromothiophene-3-carboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR17257-10G
OR17257-5G
OR17257-1G
APOSOR17257-10GEA
81.2
EUR
APOSOR17257-10G
APOSOR17257-5G
APOSOR17257-1G
5-Bromothiophene-3-carboxylic acid 97%
5-Bromothiophene-3-carboxylic acid
Formula:
C₅H₃BrO₂S MW: 207,0500 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08695074 CAS Number: 100523-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...