5-Chloro-2-nitrophenol 98%
Supplier: Apollo Scientific
|
Warning
|
OR17307-1G
OR17307-25G
OR17307-5G
APOSOR17307-1GEA
52.4
EUR
APOSOR17307-1G
APOSOR17307-25G
APOSOR17307-5G
5-Chloro-2-nitrophenol 98%
5-Chloro-2-nitrophenol
Formula:
C₆H₄ClNO₃ MW: 173,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09260849 CAS Number: 611-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...