5-Hydroxy-2-nitrobenzaldehyde 95+%
Supplier: Apollo Scientific
|
Warning
|
OR17521-5G
OR17521-10G
APOSOR17521-5GEA
21.9
EUR
APOSOR17521-5G
APOSOR17521-10G
5-Hydroxy-2-nitrobenzaldehyde 95+%
5-Hydroxy-2-nitrobenzaldehyde
Formula:
C₇H₅NO₄ MW: 167,12 g/mol Melting Pt: 165…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007332 CAS Number: 42454-06-8 EINECS: 255-832-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...