7,8-Dihydro-5(6H)-quinolinone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 7,8-Dihydroquinolin-5(6H)-one

OR17531-1G OR17531-5G OR17531-25G
APOSOR17531-1GEA 77.1 EUR
APOSOR17531-1G APOSOR17531-5G APOSOR17531-25G
7,8-Dihydro-5(6H)-quinolinone 98%
7,8-Dihydro-5(6H)-quinolinone
Formula: C₉H₉NO
MW: 147,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02181161
CAS Number: 53400-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR