1-Acetylpiperidin-4-amine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-Acetyl-4-aminopiperidine
OR17566-5G
OR17566-1G
APOSOR17566-5GEA
125
EUR
APOSOR17566-5G
APOSOR17566-1G
1-Acetylpiperidin-4-amine 97%
1-Acetylpiperidin-4-amine
Formula:
C₇H₁₄N₂O MW: 142,2000 g/mol Melting Pt: 46…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD03412449 CAS Number: 160357-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...