1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
Supplier: Apollo Scientific
OR17569-25G
OR17569-5G
APOSOR17569-25GEA
139
EUR
APOSOR17569-25G
APOSOR17569-5G
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula:
C₁₁H₁₉BN₂O₂ Melting Pt: 44…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864188 CAS Number: 847818-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...