2-Chloromalondialdehyde 96%
Supplier: Apollo Scientific
|
Warning
|
OR17624-1G
OR17624-25G
OR17624-5G
APOSOR17624-1GEA
48.6
EUR
APOSOR17624-1G
APOSOR17624-25G
APOSOR17624-5G
2-Chloromalondialdehyde 96%
2-Chloromalondialdehyde
Formula:
C₃H₃ClO₂ Melting Pt: 140…145 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01320489 CAS Number: 36437-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...