1-Amino-5-bromoisoquinoline 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromoisoquinolin-1-amine
OR17656-1G
OR17656-250MG
OR17656-50MG
APOSOR17656-1GEA
206
EUR
APOSOR17656-1G
APOSOR17656-250MG
APOSOR17656-50MG
1-Amino-5-bromoisoquinoline 95%
1-Amino-5-bromoisoquinoline
Formula:
C₉H₇BrN₂ MW: 223,0700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226920 CAS Number: 852570-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...