2-Bromo-4-iodoaniline 98%
Supplier: Apollo Scientific
|
Warning
|
OR17657-5G
OR17657-25G
OR17657-100G
APOSOR17657-5GEA
21.9
EUR
APOSOR17657-5G
APOSOR17657-25G
APOSOR17657-100G
2-Bromo-4-iodoaniline 98%
2-Bromo-4-iodoaniline
Formula:
C₆H₅BrIN MW: 297,9200 g/mol Melting Pt: 75…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780668 CAS Number: 29632-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...