2-Chloropyrimidine-4,5-diamine
Supplier: Apollo Scientific
|
Warning
|
OR17704-1G
OR17704-250MG
APOSOR17704-1GEA
32.2
EUR
APOSOR17704-1G
APOSOR17704-250MG
2-Chloropyrimidine-4,5-diamine
Formula:
C₄H₅ClN₄ Melting Pt: 230…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047355 CAS Number: 14631-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...