3-Bromostyrene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-3-vinylbenzene

OR17825-1G OR17825-25G OR17825-5G
APOSOR17825-1GEA 21.9 EUR
APOSOR17825-1G APOSOR17825-25G APOSOR17825-5G
3-Bromostyrene 97%
3-Bromostyrene
Formula: C₈H₇Br
MW: 183,05 g/mol
Boiling Pt: 74…75 °C/3mmHg
Density: 1,406 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000088
CAS Number: 2039-86-3
EINECS: 218-025-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR