2-Pyrimidinecarboxaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
OR17838-1G
OR17838-250MG
OR17838-5G
APOSOR17838-1GEA
186
EUR
APOSOR17838-1G
APOSOR17838-250MG
APOSOR17838-5G
2-Pyrimidinecarboxaldehyde 95%
2-Pyrimidinecarboxaldehyde
Formula:
C₅H₄N₂O MW: 108.1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07778416 CAS Number: 27427-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...