2,4-Dichloro-5-methylpyrimidine 98%

Supplier: Apollo Scientific
Danger

OR17853-100G OR17853-25G OR17853-5G
APOSOR17853-100GEA 60.9 EUR
APOSOR17853-100G APOSOR17853-25G APOSOR17853-5G
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
Boiling Pt: 108…109 °C/11mmHg
Melting Pt: 26…28 °C
Density: 1,39 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR