3-Bromo-5-chloropyridine-2-carbonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-5-chloropicolinonitrile
OR17893-1G
OR17893-5G
APOSOR17893-1GEA
31.9
EUR
APOSOR17893-1G
APOSOR17893-5G
3-Bromo-5-chloropyridine-2-carbonitrile 98%
3-Bromo-5-chloropyridine-2-carbonitrile
Formula:
C₆H₂BrClN₂ MW: 217,46 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12827549 CAS Number: 760207-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...