2-Amino-5-cyanopyrazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Aminopyrazine-2-carbonitrile
OR18000-5G
APOSOR18000-5GEA
151
EUR
APOSOR18000-5G
2-Amino-5-cyanopyrazine
2-Amino-5-cyanopyrazine
Formula:
C₅H₄N₄ MW: 120,1100 g/mol Melting Pt: 206…207 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08234954 CAS Number: 113305-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...