Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate 95+%

Supplier: Apollo Scientific
Warning

OR18027-250MG OR18027-1G
APOSOR18027-250MGEA 48 EUR
APOSOR18027-250MG APOSOR18027-1G
Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate 95+%
Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate
Formula: C₈H₈BrNO₃
Melting Pt: 105…107 °C
Storage Temperature: Refrigerator
CAS Number: 141761-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR