Ethyl isocyanoacetate

Supplier: Apollo Scientific
Warning

OR18110-25G OR18110-5G OR18110-100G
APOSOR18110-25GEA 190 EUR
APOSOR18110-25G APOSOR18110-5G APOSOR18110-100G
Ethyl isocyanoacetate
Ethyl isocyanoacetate
Formula: C₅H₇NO₂
Boiling Pt: 194…196 °C
Density: 1,035 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000007
CAS Number: 2999-46-4
EINECS: 221-077-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR