6-(Chloromethyl)uracil 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

OR18285-25G OR18285-5G
APOSOR18285-25GEA 133 EUR
APOSOR18285-25G APOSOR18285-5G
6-(Chloromethyl)uracil 98%
6-(Chloromethyl)uracil
Formula: C₅H₅ClN₂O₂
MW: 160,56 g/mol
Melting Pt: 256…258 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010095
CAS Number: 18592-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR