Indane
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,3-Dihydro-1H-indene, Indan
OR18290-100ML
OR18290-2.5L
OR18290-500ML
APOSOR18290-100MLEA
123
EUR
APOSOR18290-100ML
APOSOR18290-2.5L
APOSOR18290-500ML
Indane
Indane
Formula:
C₉H₁₀ MW: 118,18 g/mol Boiling Pt: 175…177 °C Melting Pt: –51 °C Density: 0,962 g/cm³ (25 °C) Flash Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003795 CAS Number: 496-11-7 EINECS: 207-814-7 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...