5-Iodouracil 97%
Supplier: Apollo Scientific
|
Warning
|
OR18306-25G
OR18306-500G
OR18306-100G
APOSOR18306-25GEA
29
EUR
APOSOR18306-25G
APOSOR18306-500G
APOSOR18306-100G
5-Iodouracil 97%
5-Iodouracil
Formula:
C₄H₃IN₂O₂ Melting Pt: 274…276 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006020 CAS Number: 696-07-1 EINECS: 211-788-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...