1,2-Diphenylethane 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Dibenzyl, Bibenzyl
OR18335-100G
OR18335-500G
OR18335-1KG
APOSOR18335-100GEA
36.4
EUR
APOSOR18335-100G
APOSOR18335-500G
APOSOR18335-1KG
1,2-Diphenylethane 98%
1,2-Diphenylethane
Formula:
(C₆H₅)CH₂CH₂(C₆H₅) MW: 182,27 g/mol Boiling Pt: 284…285 °C Melting Pt: 50…53 °C Density: 1,014 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004796 CAS Number: 103-29-7 EINECS: 203-096-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...