1,3,5-Trimethoxybenzene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
O,O,O-1,3,5-Trimethylresorcinol
OR18342-100G
OR18342-25G
OR18342-500G
APOSOR18342-100GEA
33.5
EUR
APOSOR18342-100G
APOSOR18342-25G
APOSOR18342-500G
1,3,5-Trimethoxybenzene 98%
1,3,5-Trimethoxybenzene
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol Boiling Pt: 225 °C Melting Pt: 51…53 °C Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008385 CAS Number: 621-23-8 EINECS: 210-673-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...