5,8-Dichloro-4-quinolinol

Supplier: Apollo Scientific

OR183470-1G OR183470-250MG
APOSOR183470-1GEA 524 EUR
APOSOR183470-1G APOSOR183470-250MG
5,8-Dichloro-4-quinolinol
5,8-Dichloro-4-quinolinol
Formula: C₉H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00272374
CAS Number: 53790-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR